COMH30377634 Combi-Blocks, Inc.

3-Bromo-2,6-difluoro-5-methylphenylboronic acid

  • Molecular Weight : 250.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BC-1586

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BC-1586
Availability details for
Quantity
  • Availability
  • Add to Cart