COMH30377638 Combi-Blocks, Inc.

6-(Benzyloxy)-3-{[6-(benzyloxy)-1H-indol-3-yl]methyl}-1H-indole

  • MDL number MFCD32065717
  • Cas Number : 2365418-80-8
  • Molecular Weight : 458.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4711

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4711
Availability details for
Quantity
  • Availability
  • Add to Cart