COMH3037763C Combi-Blocks, Inc.

5-Iodo-2,3-dihydro-1-benzofuran-2-carboxylic acid

  • MDL number MFCD13659384
  • Cas Number : 847949-05-7
  • Molecular Weight : 290.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4715

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4715
Availability details for
Quantity
  • Availability
  • Add to Cart