COMH30377640 Combi-Blocks, Inc.

Methyl 4-hydroxy-3-(methylamino)benzoate hydrochloride

  • MDL number MFCD32065660
  • Cas Number : 2365418-78-4
  • Molecular Weight : 217.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-6353

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-6353
Availability details for
Quantity
  • Availability
  • Add to Cart