COMH30377643 Combi-Blocks, Inc.

4-(1-Aminoethyl)phenylboronic acid hydrochloride

  • MDL number MFCD28400334
  • Cas Number : 1704073-43-7
  • Molecular Weight : 201.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BC-1596

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BC-1596
Availability details for
Quantity
  • Availability
  • Add to Cart