COMH30377677 Combi-Blocks, Inc.

Dibenzo-15-crown-5

  • MDL number MFCD00055949
  • Cas Number : 14262-60-3
  • Molecular Weight : 316.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QH-0253

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QH-0253
Availability details for
Quantity
  • Availability
  • Add to Cart