COMH303776A5 Combi-Blocks, Inc.

N-(5-Bromopyridin-3-yl)formamide

  • MDL number MFCD29950669
  • Cas Number : 1692703-11-9
  • Molecular Weight : 201.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4880

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4880
Availability details for
Quantity
  • Availability
  • Add to Cart