COMH42116C37 Combi-Blocks, Inc.

tert-Butyl N-[(tert-butoxy)carbonyl]-N-(3-isopropylphenyl)carbamate

  • MDL number MFCD32065663
  • Cas Number : 2365419-67-4
  • Molecular Weight : 335.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-6373

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-6373
Availability details for
Quantity
  • Availability
  • Add to Cart