COMH42116C38 Combi-Blocks, Inc.

4-(2,4-dinitrophenyl)-1-fluoro-2-(trifluoromethyl)benzene

  • MDL number MFCD32065664
  • Cas Number : 2365418-96-6
  • Molecular Weight : 330.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 84
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-6377

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-6377
Availability details for
Quantity
  • Availability
  • Add to Cart