COMH42116C39 Combi-Blocks, Inc.

4-Bromo-3-(dihydroxyboranyl)-2-fluorobenzoic acid

  • MDL number MFCD32065666
  • Molecular Weight : 262.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-3918

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-3918
Availability details for
Quantity
  • Availability
  • Add to Cart