COMH42116C3A Combi-Blocks, Inc.

(3-{[(tert-Butoxy)carbonyl]amino}-2-fluorophenyl)boronic acid

  • MDL number MFCD32065668
  • Cas Number : 2246573-76-0
  • Molecular Weight : 255.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-5604

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-5604
Availability details for
Quantity
  • Availability
  • Add to Cart