COMH42116C3B Combi-Blocks, Inc.

(3-{[(Benzyloxy)carbonyl]amino}-4,5-difluorophenyl)boronic acid

  • MDL number MFCD32065670
  • Cas Number : 2377609-82-8
  • Molecular Weight : 307.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-5856

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-5856
Availability details for
Quantity
  • Availability
  • Add to Cart