COMH42116C3C Combi-Blocks, Inc.

(2-Fluoro-3-formyl-6-methoxyphenyl)boronic acid

  • MDL number MFCD32065671
  • Cas Number : 2377611-41-9
  • Molecular Weight : 198.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-7001

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-7001
Availability details for
Quantity
  • Availability
  • Add to Cart