COMH42116C3E Combi-Blocks, Inc.

[3-Chloro-5-fluoro-4-(methoxycarbonyl)phenyl]boronic acid

  • MDL number MFCD32065674
  • Molecular Weight : 232.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BC-1602

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BC-1602
Availability details for
Quantity
  • Availability
  • Add to Cart