COMH42116C3F Combi-Blocks, Inc.

Bis(3-cyclopropylphenyl)boronic acid

  • MDL number MFCD32065675
  • Molecular Weight : 262.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BC-1603

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BC-1603
Availability details for
Quantity
  • Availability
  • Add to Cart