COMH42116C42 Combi-Blocks, Inc.

(5-tert-butyl-2-methylphenyl)boronic acid

  • MDL number MFCD32065678
  • Cas Number : 2377606-81-8
  • Molecular Weight : 192.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BC-1615

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BC-1615
Availability details for
Quantity
  • Availability
  • Add to Cart