COMH42116C43 Combi-Blocks, Inc.

2,3-Dichloro-4-(dihydroxyboranyl)benzoic acid

  • MDL number MFCD32065679
  • Cas Number : 2351934-06-8
  • Molecular Weight : 234.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BC-1646

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BC-1646
Availability details for
Quantity
  • Availability
  • Add to Cart