COMH42116C45 Combi-Blocks, Inc.

1-(2,2-Dimethoxyethyl)pyrazole-4-boronic acid

  • MDL number MFCD32065682
  • Cas Number : 2377611-87-3
  • Molecular Weight : 200

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (1-(2,2-dimethoxyethyl)-1H-pyrazol-4-yl)boronic acid
Inchi Code 1S/C7H13BN2O4/c1-13-7(14-2)5-10-4-6(3-9-10)8(11)12/h3-4,7,11-12H,5H2,1-2H3
Inchi Key KOIOEQAGPLXHPF-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Freezer
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-2758

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-2758
Availability details for
Quantity
  • Availability
  • Add to Cart