COMH42116C46 Combi-Blocks, Inc.

2'-(4-Methylumbelliferyl)-alpha-d-n-acetylneuraminic acid sodium salt hydrate

  • MDL number MFCD31010416
  • Cas Number : 1977472-73-3
  • Molecular Weight : 507.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-1949

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-1949
Availability details for
Quantity
  • Availability
  • Add to Cart