COMH42116C4B Combi-Blocks, Inc.

1-(Benzyloxy)-2-fluoro-3-nitrobenzene

  • MDL number MFCD32065659
  • Cas Number : 2365418-52-4
  • Molecular Weight : 247.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3269

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-3269
Availability details for
Quantity
  • Availability
  • Add to Cart