COMH42116C53 Combi-Blocks, Inc.

7-Methyl-1H-quinoxalin-2-one

  • MDL number MFCD17215508
  • Cas Number : 5762-65-2
  • Molecular Weight : 160.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-6366

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-6366
Availability details for
Quantity
  • Availability
  • Add to Cart