COMH42116C57 Combi-Blocks, Inc.

4-bromo-6-nitrobenzene-1,3-diol

  • Molecular Weight : 234.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-6371

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-6371
Availability details for
Quantity
  • Availability
  • Add to Cart