COMH42116C63 Combi-Blocks, Inc.

2-(5-Bromo-2-ethoxyphenyl)piperidine

  • MDL number MFCD11048423
  • Cas Number : 952958-95-1
  • Molecular Weight : 284.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AM-2576

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AM-2576
Availability details for
Quantity
  • Availability
  • Add to Cart