COMH42116C65 Combi-Blocks, Inc.

4-[2-(Benzyloxy)ethoxy]phenylboronic acid

  • Molecular Weight : 272.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-4026

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-4026
Availability details for
Quantity
  • Availability
  • Add to Cart