COMH42116C67 Combi-Blocks, Inc.

2-(5-Chloro-2-ethoxyphenyl)piperidine

  • MDL number MFCD05190202
  • Cas Number : 901920-26-1
  • Molecular Weight : 239.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AM-2577

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AM-2577
Availability details for
Quantity
  • Availability
  • Add to Cart