COMH42116C6A Combi-Blocks, Inc.

2-(5-Bromo-2-methoxy-3-methylphenyl)piperidine

  • MDL number MFCD32065655
  • Cas Number : 1897213-50-1
  • Molecular Weight : 284.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AM-2578

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AM-2578
Availability details for
Quantity
  • Availability
  • Add to Cart