COMH42116C72 Combi-Blocks, Inc.

9-(4-Phenylphenyl)carbazole-3-boronic acid

  • MDL number MFCD22581304
  • Cas Number : 1028648-22-7
  • Molecular Weight : 363.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BC-1598

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BC-1598
Availability details for
Quantity
  • Availability
  • Add to Cart