COMH42116C75 Combi-Blocks, Inc.

tert-Butyl 4-(4-bromo-2-methylphenyl)piperazine-1-carboxylate

  • MDL number MFCD32065656
  • Cas Number : 1613259-78-1
  • Molecular Weight : 355.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AM-2583

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AM-2583
Availability details for
Quantity
  • Availability
  • Add to Cart