COMH42116C7A Combi-Blocks, Inc.

4-fluoro-2-(4-methylpiperazin-1-yl)benzoic acid

  • MDL number MFCD26652046
  • Cas Number : 1503626-80-9
  • Molecular Weight : 238.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AM-2588

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AM-2588
Availability details for
Quantity
  • Availability
  • Add to Cart