COMH42116C80 Combi-Blocks, Inc.

1,3-dimethyl 2-(2-bromo-4-nitrophenyl)propanedioate

  • MDL number MFCD32065658
  • Cas Number : 73088-09-2
  • Molecular Weight : 332.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-2609

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-2609
Availability details for
Quantity
  • Availability
  • Add to Cart