COMH42116C8A Combi-Blocks, Inc.

3-Isopropoxy-5-methylbenzoic acid

  • MDL number MFCD27940682
  • Cas Number : 1369775-54-1
  • Molecular Weight : 194.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5653

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-5653
Availability details for
Quantity
  • Availability
  • Add to Cart