COMH42116C91 Combi-Blocks, Inc.

4-[(4-Isopropylphenyl)amino]benzoic acid

  • MDL number MFCD19156705
  • Cas Number : 1272946-08-3
  • Molecular Weight : 255.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-6065

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-6065
Availability details for
Quantity
  • Availability
  • Add to Cart