COMH42116C92 Combi-Blocks, Inc.

tert-Butyl 6-bromonaphthalene-2-carboxylate

  • MDL number MFCD25189464
  • Cas Number : 1006899-73-5
  • Molecular Weight : 307.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-6067

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-6067
Availability details for
Quantity
  • Availability
  • Add to Cart