COMH42116C98 Combi-Blocks, Inc.

4-Bromo-2-fluoro-3-nitrobenzoic acid

  • MDL number MFCD28739991
  • Cas Number : 1807025-82-6
  • Molecular Weight : 264.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?CA-6075

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?CA-6075
Availability details for
Quantity
  • Availability
  • Add to Cart