COMH42116C99 Combi-Blocks, Inc.

Selenophene-2-boronic acid

  • MDL number MFCD01319053
  • Cas Number : 35133-86-9
  • Molecular Weight : 174.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1028

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1028
Availability details for
Quantity
  • Availability
  • Add to Cart