COMH42116C9C Combi-Blocks, Inc.

2,5-Dimethylpyridine-3-boronic acid

  • MDL number MFCD07437924
  • Cas Number : 1029654-18-9
  • Molecular Weight : 151.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1414

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1414
Availability details for
Quantity
  • Availability
  • Add to Cart