COMH42116C9E Combi-Blocks, Inc.

3-(Ethoxymethyl)phenylboronic acid

  • MDL number MFCD08701750
  • Cas Number : 1107603-49-5
  • Molecular Weight : 180.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1604

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-1604
Availability details for
Quantity
  • Availability
  • Add to Cart