COMH42116CA2 Combi-Blocks, Inc.

(2-Methyl-3-carboxyphenyl)boronic acid

  • MDL number MFCD09037834
  • Cas Number : 1314264-61-3
  • Molecular Weight : 180.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-2345

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-2345
Availability details for
Quantity
  • Availability
  • Add to Cart