COMH42116CA4 Combi-Blocks, Inc.

4-Fluoropyridine-3-boronic acid

  • MDL number MFCD07368772
  • Cas Number : 860626-80-8
  • Molecular Weight : 140.91

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-2391

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-2391
Availability details for
Quantity
  • Availability
  • Add to Cart