COMH42116CA9 Combi-Blocks, Inc.

5-Bromo-2,3,4-trifluorophenylboronic acid

  • MDL number MFCD12032026
  • Cas Number : 2377611-88-4
  • Molecular Weight : 254.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-3844

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-3844
Availability details for
Quantity
  • Availability
  • Add to Cart