COMH42116CAE Combi-Blocks, Inc.

3-[(3-methoxypropoxy)methyl]phenylboronic acid

  • MDL number MFCD14687248
  • Molecular Weight : 224.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FA-4612

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FA-4612
Availability details for
Quantity
  • Availability
  • Add to Cart