COMH42116CB4 Combi-Blocks, Inc.

5-Chloro-3-methylbenzo[b]thiophen-2-ylboronic acid

  • MDL number MFCD18383106
  • Cas Number : 912331-30-7
  • Molecular Weight : 226.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FB-2232

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FB-2232
Availability details for
Quantity
  • Availability
  • Add to Cart