COMH42116CBC Combi-Blocks, Inc.

1-Chloronaphthalene-4-boronic acid, pinacol ester

  • MDL number MFCD31735828
  • Cas Number : 2128285-49-2
  • Molecular Weight : 288.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FF-2806

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FF-2806
Availability details for
Quantity
  • Availability
  • Add to Cart