COMH42116CCB Combi-Blocks, Inc.

Methyl 2-(3-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)acetate

  • MDL number MFCD18427635
  • Cas Number : 1259022-70-2
  • Molecular Weight : 294.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FF-6962

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FF-6962
Availability details for
Quantity
  • Availability
  • Add to Cart