COMH42116CCD Combi-Blocks, Inc.

N-(2-Methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)cyclopropanecarboxamide

  • MDL number MFCD18383608
  • Cas Number : 1231892-79-7
  • Molecular Weight : 301.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FF-7205

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FF-7205
Availability details for
Quantity
  • Availability
  • Add to Cart