COMH42116CCE Combi-Blocks, Inc.

1-(3-Fluorobenzyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1h-pyrazole

  • MDL number MFCD18383287
  • Cas Number : 864771-96-0
  • Molecular Weight : 302.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FF-7264

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FF-7264
Availability details for
Quantity
  • Availability
  • Add to Cart