COMH42116CCF Combi-Blocks, Inc.

4-((4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1h-pyrazol-1-yl)methyl)benzonitrile

  • MDL number MFCD18383251
  • Cas Number : 1205683-41-5
  • Molecular Weight : 309.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FF-7654

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FF-7654
Availability details for
Quantity
  • Availability
  • Add to Cart