COMH42116CD1 Combi-Blocks, Inc.

2-(Benzyloxy)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline

  • MDL number MFCD18383767
  • Cas Number : 1220715-96-7
  • Molecular Weight : 325.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FF-8180

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FF-8180
Availability details for
Quantity
  • Availability
  • Add to Cart