COMH42116CD2 Combi-Blocks, Inc.

2-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)pyrimidine-5-carboxylic acid

  • MDL number MFCD18373998
  • Molecular Weight : 326.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FF-8190

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FF-8190
Availability details for
Quantity
  • Availability
  • Add to Cart