COMH42116CD8 Combi-Blocks, Inc.

5-(tetramethyl-1,3,2-dioxaborolan-2-yl)-2-(trifluoromethoxy)pyridine

  • MDL number MFCD18733459
  • Cas Number : 1352741-13-9
  • Molecular Weight : 289.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?FM-1387

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?FM-1387
Availability details for
Quantity
  • Availability
  • Add to Cart